Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,5,6,6,7,7,7-Heptafluoro-3-iodo-1-heptanol
5,5,6,6,7,7,7-Heptafluoro-3-iodo-1-heptanol 97%
5,5,6,6,7,7,7-Heptafluoro-3-iodo-1-heptanol 97%
APOSPC450232-5G
:
Apollo Scientific
:
5,5,6,6,7,7,7-Heptafluoro-3-iodo-1-heptanol 97%
APOSPC450232-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
368.031 g/mol
:
Refrigerator
:
1993
:
3,III
:
GHS02
:
GHS07
:
89621-93-2
:
C₇H₈F₇IO
:
MFCD16619645