Order Entry
Czech Republic
ContactUsLinkComponent
3-(Trifluoromethyl)butyraldehyde 50% (w/w) in toluene
3-(Trifluoromethyl)butyraldehyde 50% (w/w) in toluene
Catalog # APOSPC450162-5G
Supplier:  Apollo Scientific
CAS Number:  
3-(Trifluoromethyl)butyraldehyde 50% (w/w) in toluene
Catalog # APOSPC450162-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :140.104 g/mol
Storage Temperature:Ambient
UN:1993
ADR:3,II
CAS Number:95853-69-3
Formula:C₅H₇F₃O
MDL Number:MFCD18074346