3-(Trifluoromethoxy)benzaldehyde oxime
Catalog # APOSPC450150-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:205.134 g/mol
Storage Temperature:Ambient
CAS Number:928063-99-4
Formula:C₈H₆F₃NO₂
MDL Number:MFCD03410420