Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
2-Ethoxy-5-fluoropyrimidine

2-Ethoxy-5-fluoropyrimidine

Supplier:   Apollo Scientific
2-Ethoxy-5-fluoropyrimidine
Supplier:  Apollo Scientific
Certificates
2-Ethoxy-5-fluoropyrimidine

2 Options Available Below

Product Details & Documents
  • CAS Number:17148-48-0
  • Formula:C₆H₇FN₂O
  • MDL Number:MFCD04972862
  • Molecular Weight:142.131 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Sekvence
    Typ balení
    Bal.
    Availability
    Price
  • 3047
    Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 3 240,00 Kč
    3 240,00 Kč
    1 * 1 g
       3 240,00 Kč

  • 3173
    Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 11 930,00 Kč
    11 930,00 Kč
    1 * 5 g
       11 930,00 Kč