
4,6-Bis(trifluoromethyl)-2(1H)-pyrimidinone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:193068-48-3
:
Formula:C₆H₂F₆N₂O
MDL Number:MFCD25978116
:232.083 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together