4,6-Bis(hydroxy)-4.6-bis(trifluoromethyl)tetrahydropyrimidine-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:281189-80-8
:
Formula:C₆H₆F₆N₂O₃
MDL Number:MFCD26408135
:268.114 g/mol
Storage Temperature:Ambient
Frequently Bought Together