Order Entry
Czech Republic
ContactUsLinkComponent
6-Fluoro-2,3-dihydro-1H-isoindol-1-one
Irritant
6-Fluoro-2,3-dihydro-1H-isoindol-1-one
Catalog # APOSPC450035-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Fluoro-2,3-dihydro-1H-isoindol-1-one
Catalog # APOSPC450035-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.138 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:340702-10-5
Formula:C₈H₆FNO
MDL Number:MFCD09701283