Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Difluoro-1-phenyl-2-(pyridin-3-yl)ethan-1-one
2,2-Difluoro-1-phenyl-2-(pyridin-3-yl)ethan-1-one
Catalog # APOSPC450014-100MG
Supplier:  Apollo Scientific
CAS Number:  
2,2-Difluoro-1-phenyl-2-(pyridin-3-yl)ethan-1-one
Catalog # APOSPC450014-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :233.213 g/mol
Storage Temperature:Ambient
CAS Number:1378261-26-7
Formula:C₁₃H₉F₂NO
MDL Number:MFCD22377945