4-[4-(Trifluoromethoxy)phenyl]-4-piperidinol hydrochloride
Catalog # APOSPC449025-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:297.701 g/mol
Storage Temperature:Ambient
CAS Number:871112-40-2
Formula:C₁₂H₁₄F₃NO₂·ClH
MDL Number:MFCD28108997