3-(4-Fluorophenoxy)azetidine hydrochloride
Catalog # APOSPC449001-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.641 g/mol
Storage Temperature:Ambient
CAS Number:63843-78-7
Formula:C₉H₁₀FNO·ClH
MDL Number:MFCD05664839