Order Entry
Czech Republic
ContactUsLinkComponent
 
3-(3-Fluorophenoxy)-azetidine hydrochloride
Catalog # APOSPC449000-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(3-Fluorophenoxy)-azetidine hydrochloride
Catalog # APOSPC449000-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.641 g/mol
Storage Temperature:Ambient
CAS Number:1236861-75-8
Formula:C₉H₁₀FNO·ClH
MDL Number:MFCD07774228