
1-Bromo-2-(trifluoromethoxy)ethane
Catalog # APOSPC4488-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.962 g/mol
Density:1.6737 g/cm³ (25 °C)
Flash Pt:110 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:1645-93-8
Formula:C₃H₄BrF₃O
MDL Number:MFCD08458097