Order Entry
Czech Republic
ContactUsLinkComponent
(2S)-2-(3,4-Difluorophenyl)oxirane, Technical Grade
Irritant
(2S)-2-(3,4-Difluorophenyl)oxirane, Technical Grade
Catalog # APOSPC448203-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(2S)-2-(3,4-Difluorophenyl)oxirane, Technical Grade
Catalog # APOSPC448203-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :156.129 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1006376-63-1
Formula:C₈H₆F₂O
MDL Number:MFCD23142999