2-Methoxy-1-(4-(trifluoromethyl)phenyl)ethanone
Catalog # APOSPC448024-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:218.172 g/mol
Storage Temperature:Ambient
CAS Number:26771-69-7
Formula:C₁₀H₉F₃O₂
MDL Number:MFCD09751273