(4-Hydroxy-3-(trifluoromethyl)phenyl)boronic acid
Catalog # APOSPC448021-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:205.927 g/mol
Storage Temperature:Ambient
CAS Number:1187874-94-7
Formula:C₇H₆BF₃O₃
MDL Number:MFCD18384143