
Trifluoropyruvic acid monohydrate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10321-14-9
:431-72-1
Formula:C₃HF₃O₃·H₂O
MDL Number:MFCD04038370
:160.049 g/mol
Melting Pt:122…125 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together