Order Entry
Czech Republic
ContactUsLinkComponent
2-(2-Chloro-4-fluorophenyl)-3-oxo-butyronitrile 95+%
2-(2-Chloro-4-fluorophenyl)-3-oxo-butyronitrile 95+%
Catalog # APOSPC446163-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Chloro-4-fluorophenyl)-3-oxo-butyronitrile 95+%
Catalog # APOSPC446163-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :211.62 g/mol
Storage Temperature:Ambient
CAS Number:1358582-97-4
Formula:C₁₀H₇ClFNO
MDL Number:MFCD17282112