5-Bromo-3-fluoro-2-methoxybenzaldehyde 95+%
Catalog # APOSPC446134-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.034 g/mol
Storage Temperature:Ambient
CAS Number:470668-70-3
Formula:C₈H₆BrFO₂
MDL Number:MFCD22035163