Order Entry
Czech Republic
ContactUsLinkComponent
2-(6-Chloro-4-trifluoromethyl-pyridin-2-ylsulphanyl)-N-ethyl-acetamide
2-(6-Chloro-4-trifluoromethyl-pyridin-2-ylsulphanyl)-N-ethyl-acetamide
Catalog # APOSPC446078-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-(6-Chloro-4-trifluoromethyl-pyridin-2-ylsulphanyl)-N-ethyl-acetamide
Catalog # APOSPC446078-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :298.712 g/mol
Storage Temperature:Ambient
CAS Number:1431555-22-4
Formula:C₁₀H₁₀ClF₃N₂OS
MDL Number:MFCD24386177