Benzoic acid N'-(6-chloro-4-trifluoromethyl-pyridin-2-yl)-N'-methyl-hydrazide
Catalog # APOSPC446077-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:329.705 g/mol
Storage Temperature:Ambient
CAS Number:1431555-17-7
Formula:C₁₄H₁₁ClF₃N₃O
MDL Number:MFCD24386176