Order Entry
Czech Republic
ContactUsLinkComponent
2-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-3,3-dimercapto-acrylonitrile
2-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-3,3-dimercapto-acrylonitrile
Catalog # APOSPC446074-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-3,3-dimercapto-acrylonitrile
Catalog # APOSPC446074-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :296.72 g/mol
Storage Temperature:Ambient
CAS Number:1431555-26-8
Formula:C₉H₄ClF₃N₂S₂
MDL Number:MFCD24386164