

2-Chloro-5-trifluoromethyl-pyridine-3-sulphonyl chloride 95%
Catalog # APOSPC446053-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
ADR:8,II
:280.052 g/mol
UN:3265
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:1208081-79-1
Formula:C₆H₂Cl₂F₃NO₂S
MDL Number:MFCD15142817