Order Entry
Czech Republic
ContactUsLinkComponent
5-Bromo-2-fluoro-3-nitropyridine 97%
5-Bromo-2-fluoro-3-nitropyridine 97%
Katalogové číslo APOSPC445050-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-2-fluoro-3-nitropyridine 97%
Katalogové číslo APOSPC445050-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.984 g/mol
Storage Temperature:Ambient
CAS Number:886372-98-1
Formula:C₅H₂BrFN₂O₂
MDL Number:MFCD07375020