Order Entry
Czech Republic
ContactUsLinkComponent
2-(5-Chloro-2-fluoro-4-nitrophenyl)-3,4-dihydronaphthalen-1(2H)-one
2-(5-Chloro-2-fluoro-4-nitrophenyl)-3,4-dihydronaphthalen-1(2H)-one
Catalog # APOSPC440019-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-(5-Chloro-2-fluoro-4-nitrophenyl)-3,4-dihydronaphthalen-1(2H)-one
Catalog # APOSPC440019-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :319.715 g/mol
Storage Temperature:Ambient
CAS Number:1451449-38-9
Formula:C₁₆H₁₁ClFNO₃
MDL Number:MFCD28154817