Order Entry
Czech Republic
ContactUsLinkComponent
 
(S)-2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-sulphide
Catalog # APOSPC440008-100MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-sulphide
Catalog # APOSPC440008-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :788.538 g/mol
Melting Pt:166…178 °C
Storage Temperature:Ambient
CAS Number:1706463-48-0
Formula:C₃₆H₁₇F₁₂O₃PS
MDL Number:MFCD28139330