
4-Fluoro-2-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:80141-94-2
:
Formula:C₉H₅F₄N
MDL Number:MFCD00061182
:203.136 g/mol
Boiling Pt:222…223 °C
Density:1.364 g/cm³ (25 °C)
Flash Pt:107.78 °C
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
Frequently Bought Together