Ethyl 7-fluoro-1,4-dihydro-6-methoxy-4-oxoquinoline-3-carboxylate
Catalog # APOSPC430609-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:265.237 g/mol
Storage Temperature:Ambient
CAS Number:622369-35-1
Formula:C₁₃H₁₂FNO₄
MDL Number:MFCD18157738