Order Entry
Czech Republic
ContactUsLinkComponent
8'-Fluoro-5'-hydroxy-spiro[cyclohexane-1,4'(1'h)-quinazolin]-2'(3'h)-one
8'-Fluoro-5'-hydroxy-spiro[cyclohexane-1,4'(1'h)-quinazolin]-2'(3'h)-one
Catalog # APOSPC430564-250MG
Supplier:  Apollo Scientific
CAS Number:  
8'-Fluoro-5'-hydroxy-spiro[cyclohexane-1,4'(1'h)-quinazolin]-2'(3'h)-one
Catalog # APOSPC430564-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :250.269 g/mol
Storage Temperature:Ambient
CAS Number:939768-67-9
Formula:C₁₃H₁₅FN₂O₂
MDL Number:MFCD14585364