Order Entry
Czech Republic
ContactUsLinkComponent
2-(Chloromethyl)-4-(trifluoromethyl)-6-(naphthalen-3-yl)pyrimidine
2-(Chloromethyl)-4-(trifluoromethyl)-6-(naphthalen-3-yl)pyrimidine
Catalog # APOSPC430562-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-(Chloromethyl)-4-(trifluoromethyl)-6-(naphthalen-3-yl)pyrimidine
Catalog # APOSPC430562-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :322.712 g/mol
Storage Temperature:Ambient
CAS Number:1206970-47-9
Formula:C₁₆H₁₀ClF₃N₂
MDL Number:MFCD14585229