Order Entry
Czech Republic
ContactUsLinkComponent
5-(4-(1,3-Dioxan-2-yl)-3-fluorophenyl)-2-chloropyridine
5-(4-(1,3-Dioxan-2-yl)-3-fluorophenyl)-2-chloropyridine
Catalog # APOSPC430551-250MG
Supplier:  Apollo Scientific
CAS Number:  
5-(4-(1,3-Dioxan-2-yl)-3-fluorophenyl)-2-chloropyridine
Catalog # APOSPC430551-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :293.721 g/mol
Storage Temperature:Ambient
CAS Number:1206969-79-0
Formula:C₁₅H₁₃ClFNO₂
MDL Number:MFCD14585365