Order Entry
Czech Republic
ContactUsLinkComponent
3-(Aminomethyl)-5-fluoro-1,3-dihydro-2H-indol-2-one
3-(Aminomethyl)-5-fluoro-1,3-dihydro-2H-indol-2-one
Catalog # APOSPC430531-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-(Aminomethyl)-5-fluoro-1,3-dihydro-2H-indol-2-one
Catalog # APOSPC430531-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :180.179 g/mol
Storage Temperature:Ambient
CAS Number:518066-41-6
Formula:C₉H₉FN₂O
MDL Number:MFCD06738904