1,1'-(Piperidine-1,3-diyl)bis(2,2,2-trifluoroethanone)
Catalog # APOSPC430326-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:277.164 g/mol
Storage Temperature:Ambient
CAS Number:1159982-57-6
Formula:C₉H₉F₆NO₂
MDL Number:MFCD11840380