Order Entry
Czech Republic
ContactUsLinkComponent
1-N-Boc-4,4-Difluoropiperidine
1-N-Boc-4,4-Difluoropiperidine
Catalog # APOSPC430323-1G
Supplier:  Apollo Scientific
CAS Number:  
1-N-Boc-4,4-Difluoropiperidine
Catalog # APOSPC430323-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.244 g/mol
Storage Temperature:Ambient
CAS Number:281652-10-6
Formula:C₁₀H₁₇F₂NO₂
MDL Number:MFCD11054110