Order Entry
Czech Republic
ContactUsLinkComponent
2-(8-Aza-bicyclo[3.2.1]octan-3-yl)acetic acid 2,2,2-trifluoroacetate
2-(8-Aza-bicyclo[3.2.1]octan-3-yl)acetic acid 2,2,2-trifluoroacetate
Catalog # APOSPC430315-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(8-Aza-bicyclo[3.2.1]octan-3-yl)acetic acid 2,2,2-trifluoroacetate
Catalog # APOSPC430315-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.244 g/mol
Storage Temperature:Ambient
CAS Number:1980044-86-7
Formula:C₉H₁₅NO₂·CF₃CO₂H
MDL Number:MFCD27978598