tert-Butyl 4-(2,5-difluorobenzoyl)piperidine-1-carboxylate
Catalog # APOSPC430280-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:325.35 g/mol
Storage Temperature:Ambient
CAS Number:1228631-03-5
Formula:C₁₇H₂₁F₂NO₃
MDL Number:MFCD16618547