Order Entry
Czech Republic
ContactUsLinkComponent
 
trans-3-Fluoro-tetrahydro-2H-pyran-4-yl methanesulphonate racemate
Catalog # APOSPC430262-1G
Supplier:  Apollo Scientific
CAS Number:  
trans-3-Fluoro-tetrahydro-2H-pyran-4-yl methanesulphonate racemate
Catalog # APOSPC430262-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.213 g/mol
Storage Temperature:Ambient
CAS Number:1864003-09-7
Formula:C₆H₁₁FO₄S
MDL Number:MFCD28348444