cis-3-Fluoro-tetrahydro-2H-pyran-4-yl methanesulphonate racemate
Catalog # APOSPC430261-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.213 g/mol
Storage Temperature:Ambient
CAS Number:1864003-53-1
Formula:C₆H₁₁FO₄S
MDL Number:MFCD28348443