Order Entry
Czech Republic
ContactUsLinkComponent
1-tert-Butyl 3-Methyl 5,5-difluoro-4-hydroxypiperidine-1,3-dicarboxylate
1-tert-Butyl 3-Methyl 5,5-difluoro-4-hydroxypiperidine-1,3-dicarboxylate
Catalog # APOSPC430260-1G
Supplier:  Apollo Scientific
CAS Number:  
1-tert-Butyl 3-Methyl 5,5-difluoro-4-hydroxypiperidine-1,3-dicarboxylate
Catalog # APOSPC430260-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :295.28 g/mol
Storage Temperature:Ambient
CAS Number:1864014-88-9
Formula:C₁₂H₁₉F₂NO₅
MDL Number:MFCD28155093