N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide
Catalog # APOSPC430254-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:348.362 g/mol
Storage Temperature:Ambient
CAS Number:182439-41-4
Formula:C₁₉H₁₉F₃N₂O
MDL Number:MFCD27978680