Order Entry
Czech Republic
ContactUsLinkComponent
N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide hydrochloride
N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide hydrochloride
Catalog # APOSPC430253-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(Piperidin-4-yl)-4'-(trifluoromethyl)biphenyl-2-carboxamide hydrochloride
Catalog # APOSPC430253-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :384.823 g/mol
Storage Temperature:Ambient
CAS Number:1838769-21-3
Formula:C₁₉H₁₉F₃N₂O·ClH
MDL Number:MFCD28016245