4-Fluoro-1-hydroxymethyl-admantane
Catalog # APOSPC430115-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.25 g/mol
Storage Temperature:Ambient
CAS Number:1283721-00-5
Formula:C₁₁H₁₇FO
MDL Number:MFCD18800685