Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloro-2-(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine ≥98%
4-Chloro-2-(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine ≥98%
Catalog # APOSPC421215-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-2-(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine ≥98%
Catalog # APOSPC421215-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :221.567 g/mol
Storage Temperature:Ambient
CAS Number:1060815-85-1
Formula:C₇H₃ClF₃N₃
MDL Number:MFCD13189354