Order Entry
Czech Republic
ContactUsLinkComponent
(R)-1-(4-Chloro-3-fluorophenyl)ethane-1,2-diol 95%
(R)-1-(4-Chloro-3-fluorophenyl)ethane-1,2-diol 95%
Catalog # APOSPC421192-500MG
Supplier:  Apollo Scientific
CAS Number:  
(R)-1-(4-Chloro-3-fluorophenyl)ethane-1,2-diol 95%
Catalog # APOSPC421192-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :190.599 g/mol
Storage Temperature:Ambient
CAS Number:1453854-84-6
Formula:C₈H₈ClFO₂
MDL Number:MFCD29077495