(E)-5-Fluoro-3,4-dihydronaphthalen-1(2H)-one oxime 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:911825-56-4
:
Formula:C₁₀H₁₀FNO
MDL Number:MFCD18642783
:179.191 g/mol
Storage Temperature:Ambient
Frequently Bought Together