5-Iodo-6-(trifluoromethyl)pyridin-2-ol 95+%
Catalog # APOSPC421074-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:288.994 g/mol
Storage Temperature:Ambient
CAS Number:1227515-14-1
Formula:C₆H₃F₃INO
MDL Number:MFCD16611365