
6-Chloro-3-iodo-2-(trifluoromethoxy)pyridine
Katalogové číslo APOSPC421024-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:323.439 g/mol
Melting Pt:33…35 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1221171-95-4
Formula:C₆H₂ClF₃INO
MDL Number:MFCD22571777