Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Chlorophenyl)-3'-fluoroacetophenone
Irritant
2-(4-Chlorophenyl)-3'-fluoroacetophenone
Catalog # APOSPC421017-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(4-Chlorophenyl)-3'-fluoroacetophenone
Catalog # APOSPC421017-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.68 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1183554-70-2
Formula:C₁₄H₁₀ClFO
MDL Number:MFCD12605188