Order Entry
Czech Republic
ContactUsLinkComponent
2-(3-Fluorophenyl)-4'-methoxyacetophenone
Irritant
2-(3-Fluorophenyl)-4'-methoxyacetophenone
Catalog # APOSPC421008-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(3-Fluorophenyl)-4'-methoxyacetophenone
Catalog # APOSPC421008-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.261 g/mol
Melting Pt:73…75 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:153529-18-1
Formula:C₁₅H₁₃FO₂
MDL Number:MFCD11545169