Order Entry
Czech Republic
ContactUsLinkComponent
2-(3,4-Difluorophenyl)-3'-fluoroacetophenone
2-(3,4-Difluorophenyl)-3'-fluoroacetophenone
Catalog # APOSPC421004-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3,4-Difluorophenyl)-3'-fluoroacetophenone
Catalog # APOSPC421004-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.216 g/mol
Storage Temperature:Ambient
CAS Number:1517619-41-8
Formula:C₁₄H₉F₃O
MDL Number:MFCD24829746