2-(3,4-Difluorophenyl)-4'-methoxyacetophenone
Catalog # APOSPC421002-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:262.251 g/mol
Storage Temperature:Ambient
CAS Number:221179-71-1
Formula:C₁₅H₁₂F₂O₂
MDL Number:MFCD12974696